missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Carbacyclin, MedChemExpress
Beskrivelse
Carbacyclin is a PGI2 analogue, acts as a prostacyclin (PGI2) receptor agonist and vasodilator, and potently inhibits platelet aggregation.
Specifications
Specifications
| Kemisk navn eller materiale | Carbacyclin |
| Fysisk form | Solid |
| CAS | 69552-46-1 |
| Farve | White |
| Mængde | 1 mg |
| Renhedsgrad noter | Research |
| Molekylær formel | C21H34O4 |
| Til brug med (applikation) | COVID-19-immunoregulation |
| Synonym | Carbaprostacyclin Carba-PGI2 |
| SMIL | O=C(O)CCC/C=C1C[C@@]2([H])C[C@@H](O)[C@H](/C=C/[C@@H](O)CCCCC)[C@@]2([H])C\1 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?