Få mere at vide
Carazolol-d6, Hydrochloride Salt, TRC
Artikelnummer. 30368002
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30368002 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30368002 Leverandør Toronto Research Chemicals Leverandørnr. TRCC1758125MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Carazolol-d6 Hydrochloride Salt
CAS 1184973-07-6
Molekylær formel C18H17D6ClN2O2
Mængde 5 mg
Synonym 1-(9H-Carbazol-4-yloxy)-3-[(1-methylethyl)amino]-2-propanol-d6 Hydrochloride, NSC 305336-d6
InChI formel InChI=1S/C18H22N2O2.ClH/c1-12(2)19-10-13(21)11-22-17-9-5-8-16-18(17)14-6-3-4-7-15(14)20-16;/h3-9,12-13,19-21H,10-11H2,1-2H3;1H/i1D3,2D3;
SMIL OC(CNC(C([2H])([2H])[2H])C([2H])([2H])[2H])COC1=CC=CC2=C1C3=C(C=CC=C3)N2.Cl
IUPAC navn 1-(9H-carbazol-4-yloxy)-3-(1,1,1,3,3,3-hexadeuteriopropan-2-ylamino)propan-2-ol;hydrochloride
Molekylvægt (g/mol) 340.88
Formel vægt 340.1825