Få mere at vide
Caramiphen Hydrochloride, Mikromol™
Beskrivelse
Caramiphen Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Caramiphen Hydrochloride |
| CAS | 125-85-9 |
| Analyt- eller komponentnavne | Caramiphen Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C18 H27 N O2 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Caramiphen Hydrochloride, Caramiphen hydrochloride (6CI), Cyclopentanecarboxylic acid, 1-phenyl-, 2-(diethylamino)ethyl ester, hydrochloride (8CI,9CI), 1-Phenylcyclopentanecarboxylic acid 2-diethylaminoethyl ester hydrochloride, 2-Diethylaminoethyl 1-phenylcyclopentanecarboxylate hydrochloride, Caramiphene hydrochloride, Caramiphenium chloride, Diethylaminoethyl 1-phenylcyclopentane-1-carboxylate hydrochloride, G 2747, Panparnit, Parpanit, Pentaphene hydrochloride, Cyclopentanecarboxylic acid, 1-phenyl-, 2-(diethylamino)ethyl ester, hydrochloride (1:1), Pentoxyverine Hydrogen Citrate Imp. B (EP), Pentoxyverine Imp. B (EP), Pentoxyverine Hydrogen Citrate Impurity B as Hydrochloride, Pentoxyverine Impurity B as Hydrochloride |
| InChI formel | InChI=1S/C18H27NO2.ClH/c1-3-19(4-2)14-15-21-17(20)18(12-8-9-13-18)16-10-6-5-7-11-16;/h5-7,10-11H,3-4,8-9,12-15H2,1-2H3;1H |
| SMIL | Cl.CCN(CC)CCOC(=O)C1(CCCC1)c2ccccc2 |
| IUPAC navn | 2-(diethylamino)ethyl 1-phenylcyclopentane-1-carboxylate;hydrochloride |
| Vis mere |