Få mere at vide
Thermo Scientific™ CaptureSelect™ LambdaXP Ligand Leakage ELISA
Shop alle Thermo Scientific produkterBeskrivelse
CaptureSelect LambdaXP Ligand Leakage ELISA is designed to detect possible leached ligand of human Fab fragments or Ig's with a lambda light chain that have been purified using CaptureSelect LambdaXP Affinity Matrix.
The LambdaXP affinity ligand is a 13-kD recombinant protein, expressed in yeast and covalently coupled to a chromatography matrix to give high chemical stability and low leakage. During the chromatography process small amounts of the affinity ligand can leach from the chromatography support and co-elute with the protein. This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect LambdaXP Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
Tekniske data
Tekniske data
| Kapacitet (metrisk) | 3 x 0.1 mL |
| Indhold og opbevaring | -20°C |
| Forsendelsestilstand | Dry ice |
| Form | Reagent Solution |
| Mængde | 1 Assay |
| Til brug med (applikation) | Affinity Ligand Leakage Quantitation Kits |
| Mål | CaptureSelect LambdaXP Ligand |
| Type | ELISA Kit |
| Tilstrækkelig til | 1 x 96-well plate |