Learn More
Thermo Scientific™ CaptureSelect™ C1-InhXL Ligand Leakage ELISA
Thermo Scientific™ CaptureSelect™ C1-InhXL Ligand Leakage ELISA is designed to detect possible leached ligand of C1-esterase inhibitor (C1Inh) purified using POROS CaptureSelect C1-InhXL Affinity Resin.
Brand: Thermo Scientific™ 810340210
Description
CaptureSelect C1-InhXL Ligand Leakage ELISA is designed to detect possible leached ligand of C1-esterase inhibitor (C1Inh) purified using POROS CaptureSelect C1-InhXL Affinity Resin. The C1-InhXL affinity ligand is a 12–15 kD recombinant single-domain antibody fragment (VHH affinity ligand) expressed in yeast and covalently coupled to a chromatography matrix for high chemical stability and low leakage. During the chromatography process small amounts of the ligand can leach from the chromatography support and co-elute with the target protein.
This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect C1-InhXL Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
Specifications
Store in freezer (-5 to -30°C). | |
Frozen Liquid | |
CaptureSelect™ C1-InhXL Affinity Ligand |
ELISA | |
10 Assays | |
Individual components |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.