missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ CaptureSelect™ Bovine IgA Affinity Matrix Product Code.: 18496658

Thermo Scientific™ CaptureSelect™ Bovine IgA Affinity Matrix

Product Code. p-7180755 Shop All Thermo Scientific Products
5 mL, 5mL
Click to view available options
Quantity:
5 mL
10 mL
Unit Size:
10mL
5mL
This item is not returnable. View return policy

Product Code. 18496658

Brand: Thermo Scientific™ 2943511005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ Bovine IgA Affinity Matrix has been designed specifically for the purification of bovine IgA from whey.

CaptureSelect Bovine IgA Affinity Matrix has been designed specifically for the purification of bovine IgA from whey.

Features of this affinity matrix include:

  • Binds monomeric, dimeric, and secretory bovine IgA
  • One-step purification from whey
  • Excellent scalability

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose-based, aldehyde-activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect Bovine IgA affinity ligand
  • Ligand coupling method: aldehyde coupling via NH2 residues of the ligand
  • Binding capacity: ∼10 g/L resin
  • Elution conditions: 0.1 M citric acid pH 3.0 or 0.1M glycine pH 3.0
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol

Specifications

Binding Capacity ∼10 g/L resin
Flow Rate 50 to 200 cm/hr. (Up to 2 bar)
Matrix Agarose-based, aldehyde-activated
Content And Storage Store at 2°C to 8°C.
Particle Size 65 ±10 μm
Form Liquid Suspension
Product Line CaptureSelect™
Quantity 5 mL
Type Affinity Matrix
Ligand Type Bovine IgA affinity ligand
Stationary Phase Anti Bovine IgA
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ CaptureSelect™ Bovine IgA Affinity Matrix >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.