Få mere at vide
Captopril EP Impurity J (as Hydrate), Mikromol™
Beskrivelse
Captopril EP Impurity J (as Hydrate)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Acetylcaptopril Hydrate |
| CAS | 801293-45-8 |
| Analyt- eller komponentnavne | Acetylcaptopril Hydrate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C11 H17 N O4 S . H2 O |
| Certificeringer/compliance | ISO 17025 |
| Synonym | (2S)-1-[(2S)-3-(Acetylsulfanyl)-2-methylpropanoyl]-pyrrolidine-2-carboxylic Acid Hydrate (Acetylcaptopril Hydrate), (2S)-1-[(2S)-3-(Acetylsulfanyl)-2-methylpropanoyl]-pyrrolidine-2-carboxylic Acid Hydrate, Captopril Imp. J (EP) as Hydrate, Acetylcaptopril Hydrate |
| InChI formel | InChI=1S/C11H17NO4S.H2O/c1-7(6-17-8(2)13)10(14)12-5-3-4-9(12)11(15)16;/h7,9H,3-6H2,1-2H3,(H,15,16);1H2/t7-,9+;/m1./s1 |
| SMIL | O.C[C@H](CSC(=O)C)C(=O)N1CCC[C@H]1C(=O)O |
| IUPAC navn | (2S)-1-[(2S)-3-acetylsulfanyl-2-methylpropanoyl]pyrrolidine-2-carboxylic acid;hydrate |
| Vis mere |