Få mere at vide
Captopril-d3, TRC
Artikelnummer. 30340925
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30340925 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30340925 Leverandør Toronto Research Chemicals Leverandørnr. TRCC1757521MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Captopril 2,5,5-d3
CAS 1356383-38-4
Molekylær formel C9 D3 H12 N O3 S
Mængde 1 mg
Synonym (2S)-1-(3-Marcapto-2-methyl-1-oxopionyl]-L-proline-2,5,5-d3, SQ-14225-d3
InChI formel InChI=1S/C9H15NO3S/c1-6(5-14)8(11)10-4-2-3-7(10)9(12)13/h6-7,14H,2-5H2,1H3,(H,12,13)/t6-,7+/m1/s1/i4D2,7D
SMIL [2H]C1([2H])CC[C@]([2H])(N1C(=O)[C@H](C)CS)C(=O)O
IUPAC navn (2S)-2,5,5-trideuterio-1-[(2S)-2-methyl-3-sulfanyl-propanoyl]pyrrolidine-2-carboxylic acid
Molekylvægt (g/mol) 220.3
Formel vægt 220.0961