Få mere at vide
Captodiame Hydrochloride, Mikromol™
Beskrivelse
Captodiame Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Captodiame Hydrochloride |
| CAS | 904-04-1 |
| Analyt- eller komponentnavne | Captodiame Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C21 H29 N S2 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Ethanamine, 2-[[[4-(butylthio)phenyl]phenylmethyl]thio]-N,N-dimethyl-, hydrochloride (1:1), Ethanamine, 2-[[[4-(butylthio)phenyl]phenylmethyl]thio]-N,N-dimethyl-, hydrochloride (9CI), Captodiame hydrochloride, Captodiamine hydrochloride, Covatine, Covatix, Suvren, Ethylamine, 2-[[p-(butylthio)-alpha-phenylbenzyl]thio]-N,N-dimethyl-, hydrochloride (6CI,7CI,8CI) |
| InChI formel | InChI=1S/C21H29NS2.ClH/c1-4-5-16-23-20-13-11-19(12-14-20)21(24-17-15-22(2)3)18-9-7-6-8-10-18;/h6-14,21H,4-5,15-17H2,1-3H3;1H |
| SMIL | Cl.CCCCSc1ccc(cc1)C(SCCN(C)C)c2ccccc2 |
| IUPAC navn | 2-[(4-butylsulfanylphenyl)-phenylmethyl]sulfanyl-N,N-dimethylethanamine;hydrochloride |
| Vis mere |