Få mere at vide
Captodiame Hydrochloride, LoGiCal
Artikelnummer. 30463149
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30463149 10 mg
1 options
Artikelnummer. 30463149 Leverandør LoGiCal Leverandørnr. LGCFOR1853.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Captodiame Hydrochloride
CAS 904-04-1
Analyt- eller komponentnavne Captodiame Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C21 H29 N S2 . Cl H
Certificeringer/compliance ISO 9001
Synonym Ethanamine, 2-[[[4-(butylthio)phenyl]phenylmethyl]thio]-N,N-dimethyl-, hydrochloride (1:1), Ethanamine, 2-[[[4-(butylthio)phenyl]phenylmethyl]thio]-N,N-dimethyl-, hydrochloride (9CI), Captodiame hydrochloride, Captodiamine hydrochloride, Covatine, Covatix, Suvren, Ethylamine, 2-[[p-(butylthio)-alpha-phenylbenzyl]thio]-N,N-dimethyl-, hydrochloride (6CI,7CI,8CI)
InChI formel InChI=1S/C21H29NS2.ClH/c1-4-5-16-23-20-13-11-19(12-14-20)21(24-17-15-22(2)3)18-9-7-6-8-10-18;/h6-14,21H,4-5,15-17H2,1-3H3;1H
Løsningstype Neat
SMIL Cl.CCCCSc1ccc(cc1)C(SCCN(C)C)c2ccccc2
IUPAC navn 2-[(4-butylsulfanylphenyl)-phenylmethyl]sulfanyl-N,N-dimethylethanamine;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 396.05
Mængde 10 mg
Formel vægt 395.1508 g/mol