missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Captan, TRC
Specifications
Specifications
| Kemisk navn eller materiale | Captan |
| CAS | 133-06-2 |
| Molekylær formel | C9 H8 Cl3 N O2 S |
| Mængde | 100 mg |
| Synonym | 4-Cyclohexene-1,2-dicarboximide, N-[(trichloromethyl)thio]- (8CI), 3a,4,7,7a-Tetrahydro-2-[(trichloromethyl)thio]-1H-isoindole-1,3(2H)-dione, Aacaptan, Amercide, Bangtan, Bangton, Buvisild K, Captab, Captadin, Captaf, Captaf 85W, Captan, Captan 4L, Captan 50W, Captan 50WP, Captan 80WDG, Captex, Crystal 50, Crystal 50 WDG, Deltan, Esso fungicide 406, Flit 406, Fungus Ban Type II, Glyodex 37-22, Hexacap, Kaptan, Kaptan 50WP, Kaptazor, Kejundan, Malipur, Merpan, Micro-Chek 12, N-Trichloromethylmercapto-4-cyclohexene-1,2-dicarboximide, N-Trichloromethylthio-3a,4,7,7a-tetrahydrophthalimide, N-Trichloromethylthio-4-cyclohexene-1,2-dicarboximide, N-[(Trichloromethyl)thio]-4-cyclohexene-1,2-dicarboximide, N-[(Trichloromethyl)thio]-δ4-tetrahydrophthalimide, N-[(Trichloromethyl)thio]tetrahydrophthalimide, NSC 36726, Neracid, Orthocide, Orthocide 406, Orthocide 50, Orthocide 7.5, Orthocide 75, Orthocide 75W, Orthocide 83, Orthocide 83RP, Orthocide S 50, Osocide, Radocaptan, Rallis captaf, SR 406, Stauffer captan, Trimegol, Ugecap, Ugecap 83, Vancide 89, Vancide 89RE, Vangard K, Venturin, Zenecal |
| InChI formel | InChI=1S/C9H8Cl3NO2S/c10-9(11,12)16-13-7(14)5-3-1-2-4-6(5)8(13)15/h1-2,5-6H,3-4H2 |
| SMIL | ClC(Cl)(Cl)SN1C(=O)C2CC=CCC2C1=O |
| IUPAC navn | 2-(trichloromethylsulfanyl)-3a,4,7,7a-tetrahydroisoindole-1,3-dione |
| Molekylvægt (g/mol) | 300.59 |
| Formel vægt | 298.9341 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?