Få mere at vide
Captan, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Captan |
| CAS | 133-06-2 |
| Molekylær formel | C9 H8 Cl3 N O2 S |
| Mængde | 1 g |
| Synonym | 4-Cyclohexene-1,2-dicarboximide, N-[(trichloromethyl)thio]- (8CI), 3a,4,7,7a-Tetrahydro-2-[(trichloromethyl)thio]-1H-isoindole-1,3(2H)-dione, Aacaptan, Amercide, Bangtan, Bangton, Buvisild K, Captab, Captadin, Captaf, Captaf 85W, Captan, Captan 4L, Captan 50W, Captan 50WP, Captan 80WDG, Captex, Crystal 50, Crystal 50 WDG, Deltan, Esso fungicide 406, Flit 406, Fungus Ban Type II, Glyodex 37-22, Hexacap, Kaptan, Kaptan 50WP, Kaptazor, Kejundan, Malipur, Merpan, Micro-Chek 12, N-Trichloromethylmercapto-4-cyclohexene-1,2-dicarboximide, N-Trichloromethylthio-3a,4,7,7a-tetrahydrophthalimide, N-Trichloromethylthio-4-cyclohexene-1,2-dicarboximide, N-[(Trichloromethyl)thio]-4-cyclohexene-1,2-dicarboximide, N-[(Trichloromethyl)thio]-δ4-tetrahydrophthalimide, N-[(Trichloromethyl)thio]tetrahydrophthalimide, NSC 36726, Neracid, Orthocide, Orthocide 406, Orthocide 50, Orthocide 7.5, Orthocide 75, Orthocide 75W, Orthocide 83, Orthocide 83RP, Orthocide S 50, Osocide, Radocaptan, Rallis captaf, SR 406, Stauffer captan, Trimegol, Ugecap, Ugecap 83, Vancide 89, Vancide 89RE, Vangard K, Venturin, Zenecal |
| InChI formel | InChI=1S/C9H8Cl3NO2S/c10-9(11,12)16-13-7(14)5-3-1-2-4-6(5)8(13)15/h1-2,5-6H,3-4H2 |
| SMIL | ClC(Cl)(Cl)SN1C(=O)C2CC=CCC2C1=O |
| IUPAC navn | 2-(trichloromethylsulfanyl)-3a,4,7,7a-tetrahydroisoindole-1,3-dione |
| Molekylvægt (g/mol) | 300.59 |
| Formel vægt | 298.9341 |