Få mere at vide
Captan, SPEX CertiPrep™
Artikelnummer. 12949464 Shop alle SPEX Certiprep produkter
Skift visning
Mængde:
1 mL
Pakningsstørrelse:
1ml
Artikelnummer. Mængde unitSize
12949464 1 mL 1ml
1 options
Artikelnummer. 12949464 Leverandør SPEX Certiprep Leverandørnr. S720
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Indeholder
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
AnalyteCAS No.Concentration
Captan133-06-21000μg/mL
Certificeringer
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Overholdelse
GHS/OSHA Complaint
Tekniske data
Kemisk navn eller materiale Captan
Emballage 2mL amber prescored glass ampule
CAS 133-06-2, 67-64-1
CAS min % 0.1, 99.9
Koncentration eller sammensætningsnoter Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molekylær formel C9H8Cl3NO2S
Til brug med (udstyr) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI nøgle LDVVMCZRFWMZSG-UHFFFAOYSA-N
SMIL C1C=CCC2C1C(=O)N(C2=O)SC(Cl)(Cl)Cl
IUPAC navn 2-(trichloromethylsulfanyl)-3a,4,7,7a-tetrahydroisoindole-1,3-dione
Opløsningsmiddel eller Matrix Acetone
Molekylvægt (g/mol) 300.578
PubChem CID 8606
Mængde 1 mL
ChEBI CHEBI:34608
Vis mere Vis mindre
Product intended for laboratory use only