missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erlab™ Captair™ Smart 392 Ductless Fume Hood
Designed with advanced carbon filtration to minimize chemical exposure.
Brand: Erlab™ A63A/AS
Description
- Sensors to detect filter breakthrough of solvents, acids or formaldehyde
- Safety back up filter in case of main filter saturation
- Continuous monitoring of air face velocity
Erlab Safety Program
- Application analysis and validation, usage certification, filter change reminders
- Connect with eGuard™ for real-time status, notifications and alerts
- Smart-Light feature for lighting up the hood
Flexibility
- Configurable filtration column
- No ductwork needed
Environment
- No dangerous chemicals are released into atmosphere
- Low energy consumption
Specifications
259 cfm | |
24.37 in. | |
52.75 in. | |
Class 1 , Type 1C, carbon filter, liquid chemical and powder handling | |
134.6 cm | |
220V 50/60Hz 105w |
61.89 cm | |
Ductless filter hood | |
31.5 in. | |
For organic vapors | |
80.01 cm |