missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals CAPSO sodium salt, 98%

Catalog Number p-4323469
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
102601-34-3
C9H12NO4S
230.26
MFCD00070063
CLEVULOKVPBHCU-VIFPVBQESA-M
capso sodium salt, sodium 3-cyclohexylamino-2-hydroxypropane-1-sulfonate, 3-cyclohexylamino-2-hydroxypropanesulfonic acid sodium salt, capso sodium, capso, 0.2m buffer solution, ph 8.5, capso, 0.2m buffer solution, ph 9.0, capso, 0.2m buffer solution, ph 9.5, sodium 3-cyclohexylamino-2-hydroxypropanesulfonate, sodium 3-cyclohexylamino-2-hydroxy-propane-1-sulfonate, potassium 3-cyclohexylamino-2-hydroxypropane-1-sulfonate
25000353
sodium;3-(cyclohexylamino)-2-hydroxypropane-1-sulfonate
O[C@@H](CNC1=CC=CC=C1)CS([O-])(=O)=O
This item is not returnable. View return policy
102601-34-3
C9H12NO4S
230.26
MFCD00070063
CLEVULOKVPBHCU-VIFPVBQESA-M
capso sodium salt, sodium 3-cyclohexylamino-2-hydroxypropane-1-sulfonate, 3-cyclohexylamino-2-hydroxypropanesulfonic acid sodium salt, capso sodium, capso, 0.2m buffer solution, ph 8.5, capso, 0.2m buffer solution, ph 9.0, capso, 0.2m buffer solution, ph 9.5, sodium 3-cyclohexylamino-2-hydroxypropanesulfonate, sodium 3-cyclohexylamino-2-hydroxy-propane-1-sulfonate, potassium 3-cyclohexylamino-2-hydroxypropane-1-sulfonate
25000353
sodium;3-(cyclohexylamino)-2-hydroxypropane-1-sulfonate
O[C@@H](CNC1=CC=CC=C1)CS([O-])(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY
Absorbance 0.050 max. (280nm, 0.1M H2O), 0.050 max. (260nm, 0.1M H20), 0.050 max. (0.1M H2O) at 280nm, 0.050 max. (280nm, 0.1M H2O), 0.050 max. (260nm
Melting Point 206°C to 211°C
Color White
pH 10.9 to 11.6 (1% aq. soln. at 25°C)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Solubility Information (10% in water) Clear colorless
Formula Weight 259.3
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material CAPSO sodium salt
Show More Show Less
compliance-icons
Product Identifier
  • CAPSO sodium salt
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'