Description
- The transparent polypropylene (PP) vials allow unobstructed viewing of the sample
- The vials are shatter-proof and may be exposed to freezing temperatures
- They feature leak-resistant screw caps that can be removed or sealed in a single turn
- Caps available in Blue, Green, Red, White and Yellow
Specifications
Specifications
| Produkttype | Screw Cap |
| Autoklaverbar | Not Autoclavable |
| Materiale | Polyethylene |
| Farve | Green |
| Sterilitet | Non-sterile |
| Til brug med (udstyr) | 6101-Tube and 6102-Tube |