Learn More
Thermo Scientific™ Capitol Vial EPA Vials
Patented flip top with permanent hinge lid
Brand: Thermo Scientific™ 04HP83HCP
Additional Details : Weight : 5.13000kg
Description
- Capacity, 118mL (4 oz.)
- 100mL EPA fill line
- High-clarity resin
- With or without sodium thiosulfate pill, string
Specifications
Clear | |
118 mL | |
Vial, Sodium thiosulfate pill, string | |
Flip Top |
Resin | |
Used to test for coliform | |
3.99 oz. |