Learn More
Trajan Scientific and Medical™ Capillary GC Column Bp5 General Purpose
A general purpose phase used for a wide range of applications
Brand: Trajan Scientific and Medical™ 054189
Additional Details : Weight : 0.01000kg
Description
- Excellent general purpose column
- Low bleed
- Non-polar
- High temperature
- Ideal for aromatics, pesticides, herbicides, drugs of abuse and hydrocarbons
Specifications
0.32 mm | |
− to 320/340°C | |
30 m |
GC Column for Environmental Analysis | |
1 μm |