missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CapG Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-00258AF700
Additional Details : Weight : 0.00970kg
Description
CapG Polyclonal specifically detects CapG in Human samples. It is validated for Western Blot, ELISA.Specifications
CapG | |
Polyclonal | |
Western Blot, ELISA | |
actin-regulatory protein CAP-G, AFCP, capping protein (actin filament), gelsolin-like, gelsolin-like capping protein, macrophage capping protein, macrophage-capping protein, MCPActin regulatory protein CAP-G | |
Produced in rabbits immunized with purified, recombinant CapG (AAH14549.1; Met1-Lys348). | |
0.1 mL | |
Cell Biology, Cellular Markers | |
822 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |