Få mere at vide
Capecitabine-d11, TRC
Artikelnummer. 30340575
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30340575 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30340575 Leverandør Toronto Research Chemicals Leverandørnr. TRCC1756525MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Capecitabine-d11
CAS 1132662-08-8
Molekylær formel C15 2H11 H11 F N3 O6
Mængde 5 mg
Synonym Cytidine, 5'-deoxy-5-fluoro-N-[(pentyl-1,1,2,2,3,3,4,4,5,5,5-d11-oxy)carbonyl]- (ACI), 5'-Deoxy-5-fluoro-N-[(pentyl-1,1,2,2,3,3,4,4,5,5,5-d11-oxy)carbonyl]cytidine (ACI), Capecitabine-d11, Capecitabine-D11 (pentyl-D11), Pentyl-d11 [1-(5-Deoxy-β-D-ribofuranosyl)-5-fluoro-2-oxo-1,2-dihydropyrimidin-4-yl]carbamate, 5'-Deoxy-5-fluoro-N-[(pentyloxy-d11)carbonyl]cytidine, [1-(5-Deoxy-β-D-ribofuranosyl)-5-fluoro-1,2-dihydro-2-oxo-4-pyrimidinyl]carbamic acid pentyl-d11 ester
InChI formel InChI=1S/C15H22FN3O6/c1-3-4-5-6-24-15(23)18-12-9(16)7-19(14(22)17-12)13-11(21)10(20)8(2)25-13/h7-8,10-11,13,20-21H,3-6H2,1-2H3,(H,17,18,22,23)/t8-,10-,11-,13-/m1/s1/i1D3,3D2,4D2,5D2,6D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])OC(=O)NC1=NC(=O)N(C=C1F)[C@@H]2O[C@H](C)[C@@H](O)[C@H]2O
IUPAC navn 1,1,2,2,3,3,4,4,5,5,5-undecadeuteriopentyl N-[1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-methyloxolan-2-yl]-5-fluoro-2-oxopyrimidin-4-yl]carbamate
Molekylvægt (g/mol) 370.42
Formel vægt 370.2183