Få mere at vide
Capecitabine-d11 (pentyl-d11), CDN
Artikelnummer. 30317646
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30317646 0.005 g
30311731 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317646 Leverandør CDN Leverandørnr. CDND75130.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Capecitabine-d11
CAS 1132662-08-8
Molekylær formel C15 2H11 H11 F N3 O6
Mængde 0.005 g
InChI formel InChI=1 S/C15H22FN3O6/c1-3-4-5-6-24-15(23)18-12-9(16)7-19(14(22)17-12)13-11(21)10(20)8(2)25-13/h7-8,10-11,13,20-21 H,3-6H2,1-2H3,(H,17,18,22,23)/t8-,10-,11-,13-/m1/s1/i1D3,3D2,4D2,5D2,6D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])OC(=O)NC1=NC(=O)N(C=C1F)[C@@H]2 O[C@H](C)[C@@H](O)[C@H]2 O
IUPAC navn 1,1,2,2,3,3,4,4,5,5,5-undecadeuteriopentyl N-[1-[(2 R,3 R,4 S,5 R)-3,4-dihydroxy-5-methyloxolan-2-yl]-5-fluoro-2-oxopyrimidin-4-yl]carbamate
Molekylvægt (g/mol) 370.42
Formel vægt 370.2183 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity