Få mere at vide
Canrenone, 98%, Thermo Scientific™
Inhibits aldosterone biosynthesis and blocks ouabain effects
Brand: Thermo Scientific Alfa Aesar J60238.03
| Mængde | 1g |
|---|
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Kemiske identifikatorer
| C22H28O3 | |
| MFCD00071735 | |
| canrenone, aldadiene, phanurane, luvion, aldadiene van, canrenone usan:inn, canrenonum inn-latin, canrenona inn-spanish, unii-78o20x9j0u, 17alpha-2-carboxyethyl-17beta-hydroxyandrosta-4,6-dien-3-one lactone | |
| (8R,9S,10R,13S,14S,17R)-10,13-dimethylspiro[2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthrene-17,5'-oxolane]-2',3-dione |
Tekniske data
| Canrenone | |
| C22H28O3 | |
| 1g | |
| canrenone, aldadiene, phanurane, luvion, aldadiene van, canrenone usan:inn, canrenonum inn-latin, canrenona inn-spanish, unii-78o20x9j0u, 17alpha-2-carboxyethyl-17beta-hydroxyandrosta-4,6-dien-3-one lactone | |
| CC12CCC(=O)C=C1C=CC3C2CCC4(C3CCC45CCC(=O)O5)C | |
| 340.463 | |
| 340.46 |
| 976-71-6 | |
| MFCD00071735 | |
| 14,1752 | |
| UJVLDDZCTMKXJK-WNHSNXHDSA-N | |
| (8R,9S,10R,13S,14S,17R)-10,13-dimethylspiro[2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthrene-17,5'-oxolane]-2',3-dione | |
| 13789 | |
| 98% |