Learn More
Poulten Selfe™ Cannon-Fenske Routine Viscometers
Cannon-Fenske viscometers, suitable for transparent liquids, with NAMAS certificate
Brand: Poulten Selfe™ 1634/08
Additional Details : Weight : 1.00000kg
Description
ASTM D445-IP71, ASTM D446.Suitable for transparent liquids, nominal overall length 250mm. Approximate sample size 7mL. Calibration data at 40°C with the constant, C, stated at 40°C and 100°C. With NAMAS certification of calibration.Specifications
Cannon-Fenske Routine Viscometer | |
D445-IP71, D446 | |
350 |
100 to 500 cSt | |
NAMAS | |
7 mL |