Få mere at vide
Cannflavin B, TRC
Artikelnummer. 30003442
Skift visning
Mængde:
2.5 mg
5 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30003442 2.5 mg 2.5mg
30008135 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30003442 Leverandør Toronto Research Chemicals Leverandørnr. TRCC1754052.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 76735-58-5
Molekylær formel C21 H22 O6
Molekylvægt (g/mol) 370.3958
Synonym 4H-1-Benzopyran-4-one, 5,7-dihydroxy-2-(4-hydroxy-3-methoxyphenyl)-6-(3-methyl-2-butenyl)- (9CI), 5,7-Dihydroxy-2-(4-hydroxy-3-methoxyphenyl)-6-(3-methyl-2-buten-1-yl)-4H-1-benzopyran-4-one, 6-Prenylchrysoeriol, Cannaflavin B, Cannflavin B, Canniflavone, Canniflavone 1, 4H-1-Benzopyran-4-one, 5,7-dihydroxy-2-(4-hydroxy-3-methoxyphenyl)-6-(3-methyl-2-buten-1-yl)-
IUPAC navn 5,7-dihydroxy-2-(4-hydroxy-3-methoxyphenyl)-6-(3-methylbut-2-enyl)-4a,8a-dihydrochromen-4-one
SMIL COc1cc(ccc1O)C2=CC(=O)C3C(O2)C=C(O)C(=C3O)CC=C(C)C
Tekniske data
Kemisk navn eller materiale Cannflavin B
Renhedsgrad noter HPLC
Mængde 2.5 mg
InChI formel InChI=1S/C21H22O6/c1-11(2)4-6-13-15(23)9-19-20(21(13)25)16(24)10-17(27-19)12-5-7-14(22)18(8-12)26-3/h4-5,7-10,19-20,22-23,25H,6H2,1-3H3
Formel vægt 370.1416
Procent renhed >95%