Learn More
Tocris Bioscience™ Candesartan
Product Code. 30001575
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001575 50 mg 50mg
30001557 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 30001575 Supplier Tocris Bioscience™ Supplier No. 4791/50
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
AT1 receptor antagonist
Angiotensin II receptor 1 (AT1) antagonist (IC50 values are 1.12 and 2.86 nM in bovine adrenal cortex and rabbit aorta, respectively). Exhibits antihypertensive effects in animal models. Also available as a prodrug, candesartan cilexetil (Cat. No. 4792).
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 139481-59-7
Molecular Formula C24H20N6O3
Specifications
Quantity 50 mg
Formula Weight 440.45
Purity >98%
Molecular Weight (g/mol) 440.463
Chemical Name or Material Candesartan