Få mere at vide
Canagliflozin Furanose Form (Mixture of Anomers), TRC
Artikelnummer. 30137904
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137904 1 mg
17964865 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137904 Leverandør Toronto Research Chemicals Leverandørnr. TRCF5940311MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2161394-94-9
Molekylær formel C24H25FO5S
Molekylvægt (g/mol) 444.52
Synonym Canagliflozin Impurity 4
SMIL Cc1ccc(cc1Cc2ccc(s2)c3ccc(F)cc3)[C@@H]4O[C@H]([C@@H](O)CO)[C@@H](O)[C@@H]4O
Tekniske data
Kemisk navn eller materiale Canagliflozin Furanose Form (Mixture of Anomers)
Mængde 1 mg
InChI formel InChI=1S/C24H25FO5S/c1-13-2-3-15(23-21(28)22(29)24(30-23)19(27)12-26)10-16(13)11-18-8-9-20(31-18)14-4-6-17(25)7-5-14/h2-10,19,21-24,26-29H,11-12H2,1H3/t19-,21-,22-,23-,24+/m0/s1
Formel vægt 444.1407