missing translation for 'onlineSavingsMsg'
Få mere at vide

Canadine, LoGiCal

Artikelnummer. 30457397
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30457397 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30457397 Leverandør LoGiCal Leverandørnr. LGCFOR3085.00

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Tetrahydroberberine (Canadine)
CAS 522-97-4
Analyt- eller komponentnavne Tetrahydroberberine (Canadine)
Forsendelsestilstand Room Temperature
Molekylær formel C20 H21 N O4
Certificeringer/compliance ISO 9001
Synonym 6H-Benzo[g]-1,3-benzodioxolo[5,6-a]quinolizine, 5,8,13,13a-tetrahydro-9,10-dimethoxy-, Berberine, 9-deoxy-16,17-dihydro- (6CI), Berbine, 9,10-dimethoxy-2,3-(methylenedioxy)- (7CI,8CI), Berberine, tetrahydro-, Canadine, NSC 36351, NSC 94918, Tetrahydroberberine, Tetrahydroumbellatine, Xanthopuccine, dl-Canadine, dl-Tetrahydroberberine, (+/-)-Canadine, (+/-)-Tetrahydroberberine
InChI formel InChI=1S/C20H21NO4/c1-22-17-4-3-12-7-16-14-9-19-18(24-11-25-19)8-13(14)5-6-21(16)10-15(12)20(17)23-2/h3-4,8-9,16H,5-7,10-11H2,1-2H3
Løsningstype Neat
SMIL COc1ccc2CC3N(CCc4cc5OCOc5cc34)Cc2c1OC
Industry Type Clinical Testing
Molekylvægt (g/mol) 339.38
Mængde 10 mg
Formel vægt 339.1471 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.