missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ cAMP Recombinant Rabbit Monoclonal Antibody (RM466)
GREENER_CHOICE

Artikelnummer. 17896731
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17896731 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17896731 Leverandør Invitrogen™ Leverandørnr. MA544556

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Recombinant Monoclonal Antibody

This antibody reacts to human IDH1(R132H).

Cyclic adenosine monophosphate (cAMP) plays a key role as an intracellular second messenger for transduction events that follow a number of extracellular signals. The G-Protein Coupled Receptors (GPCR) is the largest family of cell surface receptors. They can be activated by different ligands, such as neurotransmitters, hormones, ions, small molecules, peptides, and other physiological signaling molecules. Typically, the binding of the ligands to its receptor resulting in the activation of G-proteins, in return, activates the effector adenylyl cyclase evoking the production of cAMP. The activation of a protein kinase by cAMP results in the phosphorylation of substrate proteins. Currently successful drugs in marketing have been developed to target these receptors. Among the GPCRs, ~367 receptors are potential drug development targets, but only about 20 have been used to generate therapeutically and commercially successful drugs so far. Because the involvement of cAMP can amplify the response of the ligand binding, the second messenger cAMP has been largely employed to monitor the activation of the GPCR to facilitate the therapeutic drug discovery.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen cAMP
Applikationer ELISA
Klassifikation Recombinant Monoclonal
Klon RM466
Koncentration 1 mg/mL
Konjugeret Unconjugated
Formulering PBS with 1% BSA, 50% glycerol and 0.09% sodium azide
Gene Alias 18 kDa cationic antimicrobial protein; Antibacterial peptide FALL-39; Antibacterial peptide LL-37; CAMP; CAP18; CAP-18; Cathelicidin antimicrobial peptide; cathelin-like protein; Cathelin-related antimicrobial peptide; CLP; Cnlp; Cramp; Cyclic AMP; FALL39; FALL-39; FALL-39 peptide antibiotic; HSD26; LL37; MCLP
Værtsarter Rabbit
Immunogen KLH-conjugated cAMP.
Oprensningsmetode Protein A
Mængde 50 μg
Regulatorisk status RUO
Primær eller sekundær Primary
Målarter Chemical
Indhold og opbevaring -20°C
Produkttype Antibody
Form Liquid
Isotype IgG
Målmolekyle Chemical
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.