missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CAMKK1 (CAMKKA) Recombinant Human Protein
CAMKK1 (CAMKKA) is a serine/threonine protein kinase family member.
4375.00 DKK - 100400.00 DKK
Specifications
Accession Number | NP_115670 |
---|---|
Formulation | 50mM Tris, pH 7.5, 150mM NaCl, 0.02% Triton X-100, 0.5mM EDTA, 2mM DTT, 50 % Glycerol |
Molecular Weight (g/mol) | 59.8kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Regulatory Status | RUO - research use only |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10542605
|
Invitrogen™
PV4670 |
10μg |
4375.00 DKK
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15408673
|
Invitrogen™
PR8308A |
100μg |
21190.00 DKK
100µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15478743
|
Invitrogen™
PV4671 |
1mg |
100400.00 DKK
1mg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CAMKK1 (CAMKKA) belongs to the Ca2+/calmodulin-dependent protein kinase subfamily.
Specifications
NP_115670 | |
59.8kDa | |
RUO - research use only | |
CAMKK Kinase Family | |
Recombinant | |
Baculovirus |
50mM Tris, pH 7.5, 150mM NaCl, 0.02% Triton X-100, 0.5mM EDTA, 2mM DTT, 50 % Glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
CAMKK1 (CAMKKA) | |
Human | |
His Tag |