Learn More
Motic™ Camera Adapter, C-Mount, B3
Choose the most appropriate adapter for your camera and microscope and unlock the potential of photomicography. Motic™ Camera Adapter, C-Mount, B3 , is suitable for B3 Series Microscopes and uses the industry standard attachment for digital imaging devices dedicated to microscopes.
Brand:  Motic™ SW011364
Additional Details : Weight : 2.60000kg
 View more versions of this product
Product Code. 11858183
  • 464.00 DKK / Each
Estimated Shipment
08-07-2021
Add to basket
Description
Description
  • Connect a C-mount camera with this adapter.
  • Suitable for B3 Series Microscopes.
  • B3 Microscopes are designed for advanced laboratory work and employ ASC achromatic super contrast and plan achromatic objectives.
Specifications
Specifications
B3 Microscopes, 1/3in. Sensor Camera
C-Mount
Camera Adapter
0.5X