Få mere at vide
Cambinol, MedChemExpress
Artikelnummer. 16473138
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16473138 10 mg
16419696 50 mg
16415877 5 mg
16404436 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16473138 Leverandør Medchem Express Leverandørnr. HY100732
MedChemExpress Cambinol is a SIRT1 and SIRT2 inhibitor with IC50 values of 56 μM and 59 μM, respectively. Cambinol is a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor (exosome inhibitor).
Cambinol is a SIRT1 and SIRT2 inhibitor with IC50 values of 56 μM and 59 μM, respectively. Cambinol is a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor (exosome inhibitor).
Tekniske data
Kemisk navn eller materiale Cambinol
CAS 14513-15-6
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C21H16N2O2S
Mængde 10 mg
Opløselighedsinformation DMSO : 50 mg/mL (138.72 mM; Need ultrasonic)
SMIL O=C(C(CC1=C2C=CC=CC2=CC=C1O)=C(C3=CC=CC=C3)N4)NC4=S
Molekylvægt (g/mol) 360.43
Formel vægt 360.43
Procent renhed 98.71%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death