missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CaM Kinase II Antibody (GT4811), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP3-13508
Additional Details : Weight : 0.00970kg
Description
CaM Kinase II Monoclonal antibody specifically detects CaM Kinase II in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
CaM Kinase II | |
Monoclonal | |
Unconjugated | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
GT4811 | |
PBS | |
Recombinant protein encompassing a sequence within the center region of human CaM Kinase II. The exact sequence is proprietary. | |
100 μg | |
Apoptosis, B Cell Development and Differentiation Markers, Cell Biology, Cell Cycle and Replication, GPCR, Immunology, Neuroscience, Signal Transduction | |
816 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |