missing translation for 'onlineSavingsMsg'
Få mere at vide

Calycosin, TRC

Artikelnummer. 30344170
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30344170 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30344170 Leverandør Toronto Research Chemicals Leverandørnr. TRCC1489305MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Calycosin
CAS 20575-57-9
Molekylær formel C16 H12 O5
Mængde 5 mg
Synonym 7-Hydroxy-3-(3-hydroxy-4-methoxyphenyl)-4H-chromen-4-one, 4H-1-Benzopyran-4-one, 7-hydroxy-3-(3-hydroxy-4-methoxyphenyl)-, Isoflavone, 3',7-dihydroxy-4'-methoxy- (8CI), 3',7-Dihydroxy-4'-methoxyisoflavone, 3'-Hydroxyformononetin, 7,3'-Dihydroxy-4'-methoxyisoflavone, 7-Hydroxy-3-(3-hydroxy-4-methoxyphenyl)-4H-1-benzopyran-4-one, Calycosin, Cyclosin, Cyclosin (isoflavone)
InChI formel InChI=1S/C16H12O5/c1-20-14-5-2-9(6-13(14)18)12-8-21-15-7-10(17)3-4-11(15)16(12)19/h2-8,17-18H,1H3
SMIL COc1ccc(cc1O)C2=COc3cc(O)ccc3C2=O
IUPAC navn 7-hydroxy-3-(3-hydroxy-4-methoxyphenyl)chromen-4-one
Molekylvægt (g/mol) 284.26
Formel vægt 284.0685
Produkttitel
Vælg et problem

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.