missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ Calsequestrin Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA585406

Produktkode 16352345

  • 4000.00 DKK / 100µL

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolitik

Beskrivelse

Beskrivelse

Keep as concentrated solution. Predicted reactivity: Mouse (96%), Rat (96%), Pig (96%), Rabbit (96%), Bovine (94%). Positive Control: HepG2, mouse muscle, rat muscle. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The sarcoplasmic reticulum (SR) is, in part, responsible for maintaining the level of intracellular calcium in cardiac and skeletal muscle by storing and releasing calcium. Several intralumenal SR calcium binding proteins have been identified, the most prominent of these is calsequestrin. Calsequstrin is a calcium binding protein known to sequester calcium accumulated in the sarcoplasmic reticulum of muscle cells during relaxation and is found discretely localized to the junctional and corbular (terminal cisternae) SR. Calsequestrin functions to localize calcium near the junctional face of the terminal cisternae from which calcium can be released into the cytosol via the ryanodine receptor. This protein is highly acidic and has a large capacity and moderate to low affinity for calcium.
TRUSTED_SUSTAINABILITY
Tekniske data

Tekniske data

Calsequestrin
Polyclonal
Unconjugated
CASQ1
Aspartactin; calmitin; calmitine; calsequestrin; calsequestrin 1; calsequestrin 1 (fast-twitch, skeletal muscle); calsequestrin, skeletal muscle isoform; Calsequestrin-1; calsquestrin 1; CASQ; Casq1; CSQ; CSQ1; CSQ-1; Laminin-binding protein; PDIB1; sCSQ; skeletal muscle calsequestrin 1; VMCQA
Rabbit
Antigen affinity chromatography
RUO
12372, 686019, 844
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.79 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
O09165, P19633, P31415
CASQ1
Recombinant protein encompassing a sequence within the center region of human Calsequestrin 1. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Produktforslag

Produktforslag

Videoer
SDS
Dokumenter

Dokumenter

Certifikater
Kampagner

Kampagner

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.