missing translation for 'onlineSavingsMsg'
Learn More

Calnexin (Endoplasmic Reticulum Marker) Mouse, Clone: CANX/1543, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 821-MSM3-P0

 View more versions of this product

Product Code. 15961128

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Calnexin (Endoplasmic Reticulum Marker)
Cytoplasmic
CANX/1543
Unconjugated
Mouse
90kDa
Protein A/G purified from Bioreactor concentrate.
RUO
821
IgG1 Îş
ELISA, Western Blot, Immunohistochemistry
Monoclonal
200ÎĽg/mL
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Recombinant N-terminal fragment of human Calnexin protein (apprx. aa 1-300)
HeLa, MCF-7 or U2OS cells. Kidney or small intestine.
20 ÎĽg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calnexin (Endoplasmic Reticulum Marker) Mouse, Clone: CANX/1543, enQuireBio™ > 20 μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.