missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CALML5 Antibody (119), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89981AF405
Additional Details : Weight : 0.00970kg
Description
CALML5 Monoclonal specifically detects CALML5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CALML5 | |
Monoclonal | |
Alexa Fluor 405 | |
calmodulin-like 5, calmodulin-like protein 5, CLSPCalmodulin-like skin protein | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CALML5 (AAH39172.1; Met1-Glu146). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
119 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
51806 | |
Store at 4°C in the dark. | |
IgG |