Få mere at vide
Calindol-13C,d2 Hydrochloride, TRC
Artikelnummer. 30329353
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30329353 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30329353 Leverandør Toronto Research Chemicals Leverandørnr. TRCC1487021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Calindol-13C,D2 Hydrochloride
CAS 1217828-76-6
Molekylær formel C2013CH19D2ClN2
Mængde 1 mg
InChI formel InChI=1S/C21H20N2.ClH/c1-15(19-11-6-9-16-7-2-4-10-20(16)19)22-14-18-13-17-8-3-5-12-21(17)23-18;/h2-13,15,22-23H,14H2,1H3;1H/t15-;/m1./s1/i14+1D2;
SMIL Cl.[2H][13C]([2H])(N[C@H](C)c1cccc2ccccc12)c3cc4ccccc4[nH]3
IUPAC navn (1R)-N-[dideuterio(1H-indol-2-yl)(113C)methyl]-1-naphthalen-1-ylethanamine;hydrochloride
Molekylvægt (g/mol) 339.86
Formel vægt 339.1552