Få mere at vide
California Pesticide Class 1 Mixture cis and trans Chlordane 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Aldicarb∣Carbofuran∣Chlorfenapyr∣Chlorpyrifos∣Cis-Chlordane (Alpha Isomer)∣Coumaphos∣Daminozide∣Dichlorvos∣Dimethoate∣Ethoprophos∣Etofenprox∣Fenoxycarb∣Fipronil∣Imazalil∣Methiocarb∣Mevinphos∣Paclobutrazol∣Parathion-methyl∣Propoxur∣Spiroxamine∣Thiacloprid∣Trans-Chlordane (Gamma Isomer) |
| CAS | 116-06-3, 1563-66-2, 122453-73-0, 2921-88-2, 5103-71-9, 56-72-4 |
| Analyt- eller komponentnavne | Aldicarb∣Carbofuran∣Chlorfenapyr∣Chlorpyrifos∣Cis-Chlordane (Alpha Isomer)∣Coumaphos∣Daminozide∣Dichlorvos∣Dimethoate∣Ethoprophos∣Etofenprox∣Fenoxycarb∣Fipronil∣Imazalil∣Methiocarb∣Mevinphos∣Paclobutrazol∣Parathion-methyl∣Propoxur∣Spiroxamine∣Thiacloprid∣Trans-Chlordane (Gamma Isomer) |
| Forsendelsestilstand | Room Temperature |
| Certificeringer/compliance | ISO 17034 |
| Opløsningsmiddel eller Matrix | Acetonitrile |
| Mængde | 1 ml |
| Koncentration eller sammensætning (efter analyt eller komponenter) | Aldicarb 100 μg/mL∣Carbofuran 100 μg/mL∣Chlorfenapyr 100 μg/mL∣Chlorpyrifos 100 μg/mL∣Cis-Chlordane (Alpha Isomer) 100 μg/mL∣Coumaphos 100 μg/mL∣Daminozide 100 μg/mL∣Dichlorvos 100 μg/mL∣Dimethoate 100 μg/mL∣Ethoprophos 100 μg/mL∣Etofenprox 100 μg/mL∣Fenoxycarb 100 μg/mL∣Fipronil 100 μg/mL∣Imazalil 100 μg/mL∣Methiocarb 100 μg/mL∣Mevinphos 100 μg/mL∣Paclobutrazol 100 μg/mL∣Parathion-methyl 100 μg/mL∣Propoxur 100 μg/mL∣Spiroxamine 100 μg/mL∣Thiacloprid 100 μg/mL∣Trans-Chlordane (Gamma Isomer) 100 μg/mL |