Learn More
HUBER Calibration Bend
Calibration Bend, accessories for Unistats
Brand: HUBER 9925
Additional Details : Weight : 0.53000kg
Description
Calibration bend with sensor pocketSpecifications
6 dia. x 160 mm Internal Sensor | |
Calibration Bend | |
M38 x 1.5 |