missing translation for 'onlineSavingsMsg'
Få mere at vide

Calhex 231, TRC

Artikelnummer. 30339976
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30339976 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30339976 Leverandør Toronto Research Chemicals Leverandørnr. TRCC14800025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Calhex 231
CAS 652973-93-8
Molekylær formel C25 H27 Cl N2 O
Mængde 25 mg
InChI formel InChI=1S/C25H27ClN2O/c1-17(21-10-6-8-18-7-2-3-9-22(18)21)27-23-11-4-5-12-24(23)28-25(29)19-13-15-20(26)16-14-19/h2-3,6-10,13-17,23-24,27H,4-5,11-12H2,1H3,(H,28,29)/t17-,23+,24+/m1/s1
SMIL C[C@@H](N[C@H]1CCCC[C@@H]1NC(=O)c2ccc(Cl)cc2)c3cccc4ccccc34
IUPAC navn 4-chloro-N-[(1S,2S)-2-[[(1R)-1-naphthalen-1-ylethyl]amino]cyclohexyl]benzamide
Molekylvægt (g/mol) 406.95
Formel vægt 406.1812
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.