Få mere at vide
Tocris Bioscience™ Calhex 231 hydrochloride
Artikelnummer. 18045252
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18045252 10 mg
18035122 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18045252 Leverandør Tocris Bioscience™ Leverandørnr. 4387/10
Negative allosteric modulator of calcium-sensing receptor (CaSR)
Negative allosteric modulator of the calcium sensing receptor (CaSR). Blocks increases in [3H]inositol phosphate levels elicited by wild-type hCasR activation (IC50= 0.39 μM in transiently transfected HEK293 cells).
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 652973-93-8
Molekylær formel C25H27ClN2O
Tekniske data
Kemisk navn eller materiale Calhex 231 hydrochloride
Mængde 10 mg
Formel vægt 443.41
Renhed >98%
Molekylvægt (g/mol) 406.954