Learn More
Calcium Sulfate, 99%, -325 mesh, Honeywell Fluka™
Catalog Number 15642360
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
7778-18-9
CaO4S
136.13
MFCD00010912
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
calcium sulfate, drierite, gypsum, sulfuric acid, calcium salt 1:1, karstenite, calcium sulphate, anhydrous gypsum, plaster of paris, anhydrous calcium sulfate, calcium sulfate, anhydrous
24497
CHEBI:31346
[Ca++].[O-]S([O-])(=O)=O
7778-18-9
CaO4S
136.13
MFCD00010912
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
calcium sulfate, drierite, gypsum, sulfuric acid, calcium salt 1:1, karstenite, calcium sulphate, anhydrous gypsum, plaster of paris, anhydrous calcium sulfate, calcium sulfate, anhydrous
24497
CHEBI:31346
[Ca++].[O-]S([O-])(=O)=O
−325 mesh, 99%
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Color White
Quantity 500 g
Linear Formula CaSO4
UN Number NONH for all modes of transport
IUPAC Name calcium sulfate
Formula Weight 136.14g/mol
Percent Purity 99%
Packaging Poly Bottle
Appearance Powder
Density 2.960g/cm3
Chemical Name or Material Calcium Sulfate