missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Phosphate Dibasic, MP Biomedicals
Click to view available options
Quantity:
1 kg
Specifications
Quantity | 1 kg |
Formula Weight | 136.1 |
Chemical Name or Material | Calcium Phosphate Dibasic |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'