missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Nitrate Tetrahydrate, ≥99.0%, Honeywell Fluka™
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Melting Point | 42°C |
Color | Colorless |
pH | 4.0 to 7.0 (at 20°C) |
Physical Form | Crystalline |
Quantity | 500 g |
Linear Formula | Ca(NO3)2 · 4H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 236.15g/mol |
Percent Purity | ≥99.0% |
Grade | FCC |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'