missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcitonin Antibody (CALCA/3310), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14041H
Additional Details : Weight : 0.00970kg
Description
Calcitonin Monoclonal antibody specifically detects Calcitonin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Calcitonin | |
Monoclonal | |
HRP | |
ATP2B2plasma membrane calcium ATPase, ATPase, Ca++ transporting, plasma membrane 4, DKFZp686G08106, DKFZp686M088, EC 3.6.3, EC 3.6.3.8, Matrix-remodeling-associated protein 1, matrix-remodelling associated 1, MXRA1, Plasma membrane calcium ATPase isoform 4, plasma membrane calcium pump, Plasma membrane calcium pump isoform 4, plasma membrane calcium-transporting ATPase 4, PMCA4b, PMCA4PMCA4x, sarcolemmal calcium pump | |
Recombinant fragment of human Calcitonin protein (around aa 3-116) (exact sequence is proprietary) (Uniprot: P01258 ) | |
0.1 mL | |
Cancer, Cellular Markers, Neuroscience | |
796 | |
Store at 4°C in the dark. | |
IgG2b κ |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
CALCA/3310 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |