Få mere at vide
Calcitetrol, MedChemExpress
Artikelnummer. 16478917
Encompass_Preferred
Skift visning
Mængde:
1 mg
5 mg
Pakningsstørrelse:
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16478917 5 mg -
16471479 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16478917 Leverandør Medchem Express Leverandørnr. HY15157
MedChemExpress Calcitetrol(1(alpha), 24, 25-Trihydroxy VD3) is the hormonally active form of vitamin D with three hydroxyl groups.
Calcitetrol(1(alpha), 24, 25-Trihydroxy VD3) is the hormonally active form of vitamin D with three hydroxyl groups.
Tekniske data
Kemisk navn eller materiale Calcitetrol
CAS 72203-93-1
Farve White
Renhedsgrad noter Research
Molekylær formel C27H44O4
Mængde 5 mg
Synonym 1α, 24, 25-Trihydroxy VD3
Opløselighedsinformation DMSO : 100 mg/mL (231.14 mM; Need ultrasonic)
SMIL O[C@@H](C[C@H](O)C/1=C)CC1=C\C=C2[C@@](CC[C@@H]3[C@@H](CCC(O)C(C)(O)C)C)([H])[C@]3(C)CCC\2
Molekylvægt (g/mol) 432.64
Formel vægt 432.64
Procent renhed 98.41%
Grad Research
Fysisk form Solid
Til brug med (applikation) Metabolism-sugar/lipid metabolism