missing translation for 'onlineSavingsMsg'
Få mere at vide

Tocris Bioscience™ Calcipotriol
GREENER_CHOICE

Artikelnummer. 18094602
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18094602 10 mg
18064182 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18094602 Leverandør Tocris Bioscience™ Leverandørnr. 2700/10

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Vitamin D3 analog

Calcipotriol is a vitamin D3 analog that displays minimal effects on calcium homeostasis. Regulates cell differentiation and proliferation; exhibits antiproliferative activity against human HL-60, HL60/MX2, keratinocytes, MCF-7, T47D, SCC-25, and mouse WEHI-3 cancer cell lines. Calcipotriol suppresses skin cancer development in mice in a thymic stromal lymphopoietin (TSLP)-dependent manner. Calcipotriol promotes the differentiation of keratinocytes. In human T cells and in psoriatic skin, Calcipotriol increases IL-10 secretion, induces secretion of type 2 cytokines by T cells and modulates the differentiation of dendritic cells and T cells.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112965-21-6
Molekylær formel C27H40O3

Specifications

Kemisk navn eller materiale Calcipotriol
Mængde 10 mg
Formel vægt 412.6
Renhed >98%
Molekylvægt (g/mol) 412.61
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.