Få mere at vide
MedChemExpress Calcein Blue
Tekniske data
Tekniske data
| Indhold og opbevaring | 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light) |
| Produkttype | Dye Reagents |
| Mængde | 25 mg |
| Forsendelsestilstand | Room temperature |
| Opløselighed | DMSO : 6 mg/mL (18.68 mM; Need ultrasonic and warming) ∣ H2O : 1 mg/mL (3.11 mM; ultrasonic and warming and heat to 80°C) |
| Molekylvægt (g/mol) | 321.28 g/mol |
| CAS | 54375-47-2 |