Få mere at vide
Caffeine D9 (trimethyl D9) 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Artikelnummer. 30324239
Skift visning
Mængde:
1 mL
Artikelnummer. Mængde
30324239 1 mL
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324239 Leverandør Dr. Ehrenstorfer Leverandørnr. DREA11693040AL100
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Caffeine-d9
CAS 72238-85-8
Opløsningsmiddel eller Matrix Acetonitrile
Molekylær formel C8 2H9 H N4 O2
Mængde 1 mL
Synonym 1H-Purine-2,6-dione, 3,7-dihydro-1,3,7-tri(methyl-d3)-, 3,7-Dihydro-1,3,7-tri(methyl-d3)-1H-purine-2,6-dione, Caffeine-d9
InChI formel InChI=1S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3/i1D3,2D3,3D3
SMIL [2H]C([2H])([2H])N1C(=O)N(c2ncn(c2C1=O)C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 1,3,7-tris(trideuteriomethyl)purine-2,6-dione
Molekylvægt (g/mol) 203.25
Formel vægt 203.1369 g/mol