Få mere at vide
Caffeine-D10, TRC
Artikelnummer. 30328992
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30328992 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30328992 Leverandør Toronto Research Chemicals Leverandørnr. TRCC12203010MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Caffeine-D10
CAS 1220356-33-1
Molekylær formel C8D10N4O2
Mængde 10 mg
Synonym 3,7-Dihydro-1,3,7-tri(methyl-d3)-1H-purine-2,6-dione-8-d
InChI formel InChI=1S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3/i1D3,2D3,3D3,4D
SMIL [2H]C(N1C([2H])([2H])[2H])=NC(N2C([2H])([2H])[2H])=C1C(N(C([2H])([2H])[2H])C2=O)=O
IUPAC navn 8-deuterio-1,3,7-tris(trideuteriomethyl)purine-2,6-dione
Molekylvægt (g/mol) 204.25